Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGAP22 SIRNA Assays ›
Gene Symbol:
ARHGAP22
Gene Name:
Rho GTPase activating protein 22
Gene Aliases:
RhoGAP2, RhoGap22
Chromosome Location:
Chr.10: 48429831 - 48656265 on Build GRCh38
UniGene ID:
Hs.655672
Species:
Human
siRNA ID
HSS126895
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGAP22
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256024.1
NP_001242953.1
8
1266
NM_001256025.2
NP_001242954.1
8
1132
NM_001256026.1
NP_001242955.1
6
772
NM_021226.3
NP_067049.2
8
1218
NR_045675.1
9
1717
XM_005270014.3
XP_005270071.1
6
676
XM_011540002.1
XP_011538304.1
9
1817
XM_011540003.1
XP_011538305.1
9
1119
XM_011540004.1
XP_011538306.1
9
1769
XM_011540005.1
XP_011538307.1
6
820
XM_011540006.1
XP_011538308.1
8
1640
XM_011540007.2
XP_011538309.1
6
5024
XM_011540011.2
XP_011538313.1
5
499
XM_011540012.2
XP_011538314.1
6
1177
XM_011540013.2
XP_011538315.1
9
1817
XM_011540015.2
XP_011538317.1
9
1817
XM_017016470.1
XP_016871959.1
7
1086
XM_017016471.1
XP_016871960.1
6
1091
XM_017016472.1
XP_016871961.1
6
664
XM_017016473.1
XP_016871962.1
9
1769
XM_017016474.1
XP_016871963.1
8
1215
XM_017016475.1
XP_016871964.1
7
1086
XM_017016476.1
XP_016871965.1
8
1215
XM_017016477.1
XP_016871966.1
8
1163
GenBank mRNA
AK055037.1
-
7
811
AK055937.1
-
9
1717
AK127586.1
-
6
772
AK293579.1
-
8
1018
AY324801.1
-
8
1180
BC047096.1
-
8
1218
BC126444.1
-
8
1087
8992160
BC136319.1
-
8
1087
9020306
EF560749.1
-
8
1190
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
signal transduction
cell differentiation
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
GTPase activator activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline