Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL22RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021258.3 | NP_067081.2 | 5 | 799 | ||
XM_011541882.1 | XP_011540184.1 | 6 | 748 | |||
GenBank mRNA | AF286095.1 | - | 5 | 619 | ||
AK226001.1 | - | 5 | 633 | |||
AK292204.1 | - | 5 | 694 | |||
AK304454.1 | - | 3 | 424 | |||
AK308522.1 | - | 6 | 697 | |||
AK313971.1 | - | 5 | 799 | |||
BC029273.1 | - | 5 | 635 | 4347189 |