Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021229.3 | NP_067052.2 | 2 | 991 | ||
GenBank mRNA | AF119916.3 | - | 3 | 528 | ||
AF278532.1 | - | 2 | 993 | |||
AF297711.1 | - | 2 | 843 | |||
AK024691.1 | - | 1 | 67 | |||
AK226070.1 | - | 2 | 848 | |||
AK290591.1 | - | 2 | 528 | |||
AK315796.1 | - | 2 | 850 | |||
AL833767.1 | - | 1 | 395 | |||
AY330211.1 | - | 2 | 542 | |||
BC013591.1 | - | 1 | 66 | |||
BC136798.1 | - | 2 | 966 | 9020788 | ||
BC136799.1 | - | 2 | 966 | 9020789 |