Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021619.2 | NP_067632.2 | 3, 4 | 633 | ||
GenBank mRNA | AY004252.2 | - | 3, 4 | 633 |