Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021626.2 | NP_067639.1 | 8 | 766 | ||
XM_005257557.2 | XP_005257614.1 | 8 | 748 | |||
GenBank mRNA | AF113214.1 | - | 8 | 691 | ||
AF265441.1 | - | 4 | 235 | |||
AF282618.1 | - | 8 | 745 | |||
AK027373.1 | - | 8 | 739 | |||
AK092928.1 | - | 4 | 1567 | |||
AK298161.1 | - | 8 | 685 | |||
AY358559.1 | - | 8 | 746 | |||
BC072405.1 | - | 8 | 716 | 6137668 |