Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021627.2 | NP_067640.2 | 14, 15 | 1689 | ||
XM_005247690.2 | XP_005247747.2 | 13, 14 | 1572 | |||
XM_005247691.2 | XP_005247748.1 | 14, 15 | 1601 | |||
XM_017006973.1 | XP_016862462.1 | 13, 14 | 1469 | |||
GenBank mRNA | AF151697.2 | - | 14, 15 | 1692 | ||
AK027599.1 | - | 14, 15 | 1630 | |||
AK074357.1 | - | 9, 10 | 1060 | |||
AK296744.1 | - | 11, 12 | 1234 | |||
AK296772.1 | - | 13, 14 | 1570 | |||
AK298628.1 | - | 14, 15 | 1516 | |||
AK304402.1 | - | 14, 15 | 1758 | |||
AL834380.1 | - | 14, 15 | 1513 | |||
BC040609.1 | - | 14, 15 | 1575 | 5163937 |