Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RXFP1 SIRNA Assays ›
Gene Symbol:
RXFP1
Gene Name:
relaxin/insulin like family peptide receptor 1
Gene Aliases:
LGR7, RXFPR1
Chromosome Location:
Chr.4: 158521711 - 158653372 on Build GRCh38
UniGene ID:
Hs.591686
Species:
Human
siRNA ID
HSS126997
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RXFP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001253727.1
NP_001240656.1
15, 16
1690
NM_001253728.1
NP_001240657.1
14, 15
1510
NM_001253729.1
NP_001240658.1
13, 14
1465
NM_001253730.1
NP_001240659.1
14, 15
1531
NM_001253732.1
NP_001240661.1
16, 17
1693
NM_001253733.1
NP_001240662.1
15, 16
1621
NM_021634.3
NP_067647.2
15, 16
1609
NR_045579.1
15, 16
2226
NR_045580.1
15, 16
1662
NR_045581.1
15, 16
1633
NR_045582.1
13, 14
1570
NR_045583.1
14, 15
1549
XM_011532174.1
XP_011530476.1
15, 16
1656
XM_011532175.1
XP_011530477.1
14, 15
1587
XM_011532176.1
XP_011530478.1
14, 15
1509
XM_017008517.1
XP_016864006.1
14, 15
1584
XM_017008518.1
XP_016864007.1
15, 16
1578
XM_017008519.1
XP_016864008.1
16, 17
1623
XM_017008520.1
XP_016864009.1
15, 16
1712
XM_017008521.1
XP_016864010.1
16, 17
1542
XM_017008522.1
XP_016864011.1
15, 16
1627
XM_017008526.1
XP_016864015.1
10, 11
1080
GenBank mRNA
AB208943.1
-
16, 17
1431
AF190500.1
-
15, 16
1346
AK122647.1
-
17, 18
1650
AK294694.1
-
15, 16
1454
AK295040.1
-
15, 16
2049
AK300379.1
-
14, 15
1405
AK307889.1
-
14, 15
1448
AK312470.1
-
15, 16
1453
AY899849.1
-
13, 14
1202
AY899850.1
-
14, 15
1247
BC112142.1
-
15, 16
1428
8327610
BC113617.1
-
15, 16
1431
8322669
BC143556.1
-
15, 16
1451
BC143558.1
-
15, 16
1484
BC143559.1
-
14, 15
1353
BX647975.1
-
15, 16
1547
BX647980.1
-
15, 16
1547
BX647985.1
-
15, 16
1576
DA308373.1
-
1, 2
227
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9xzrh:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline