Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM3A SIRNA Assays ›
Gene Symbol:
FAM3A
Gene Name:
family with sequence similarity 3 member A
Gene Aliases:
2.19, DLD, DXS560S, XAP-7
Chromosome Location:
Chr.X: 154506159 - 154516249 on Build GRCh38
UniGene ID:
Hs.289108
Species:
Human
siRNA ID
HSS127017
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM3A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001171132.2
NP_001164603.1
8, 9
849
NM_001171133.2
NP_001164604.1
7, 8
735
NM_001171134.2
NP_001164605.1
6, 7
895
NM_001282311.1
NP_001269240.1
8, 9
1044
NM_001282312.1
NP_001269241.1
8, 9
790
NM_021806.3
NP_068578.2
7, 8
946
XM_005274714.3
XP_005274771.1
9, 10
915
XM_005274716.3
XP_005274773.1
9, 10
894
XM_005277879.4
XP_005277936.1
7, 8
804
XM_006724831.2
XP_006724894.1
9, 10
918
XM_006724832.3
XP_006724895.1
8, 9
879
XM_006724833.2
XP_006724896.1
8, 9
843
XM_006724834.2
XP_006724897.1
8, 9
1031
XM_011531185.2
XP_011529487.1
6, 7
476
XM_017029699.1
XP_016885188.1
7, 8
1191
XM_017029700.1
XP_016885189.1
7, 8
1166
XM_017029701.1
XP_016885190.1
8, 9
1069
XM_017029702.1
XP_016885191.1
7, 8
682
GenBank mRNA
AK024601.1
-
8, 9
807
AK057945.1
-
8, 9
664
AK124890.1
-
4, 5
3137
AK294116.1
-
6, 7
858
AK314682.1
-
7, 8
864
BC002934.1
-
8, 9
948
BC008912.2
-
7, 8
597
3639043
CX867071.1
-
7, 8
653
X87193.1
-
7, 8
753
Back To Top
More Information
Panther Classification:
Molecular Function -
antimicrobial response protein
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline