Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDA2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199687.2 | NP_001186616.1 | 6 | 738 | ||
NM_001242310.1 | NP_001229239.1 | 6 | 703 | |||
NM_001324199.1 | NP_001311128.1 | 4 | 384 | |||
NM_001324201.1 | NP_001311130.1 | 5 | 658 | |||
NM_001324206.1 | NP_001311135.1 | 6 | 746 | |||
NM_021783.4 | NP_068555.1 | 6 | 740 | |||
NR_136726.1 | 6 | 647 | ||||
NR_136727.1 | 6 | 649 | ||||
XM_011531000.1 | XP_011529302.1 | 7 | 753 | |||
XM_011531001.1 | XP_011529303.1 | 7 | 801 | |||
XM_011531003.2 | XP_011529305.1 | 5 | 553 | |||
XM_011531005.2 | XP_011529307.1 | 4 | 387 | |||
XM_017029703.1 | XP_016885192.1 | 5 | 650 | |||
XM_017029705.1 | XP_016885194.1 | 5 | 528 | |||
XM_017029707.1 | XP_016885196.1 | 5 | 437 | |||
GenBank mRNA | AF298812.1 | - | 5 | 630 | ||
AK296264.1 | - | 6 | 599 | |||
AK299018.1 | - | 4 | 340 | |||
AK314881.1 | - | 6 | 706 | |||
AY152724.1 | - | 6 | 693 | |||
AY358735.1 | - | 5 | 639 | |||
AY358736.1 | - | 5 | 633 | |||
BC034919.1 | - | 6 | 687 | 5434545 | ||
DC368837.1 | - | 4 | 340 |