Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037126.1 | NP_001032203.1 | 8 | 1305 | ||
NM_021807.3 | NP_068579.3 | 8 | 1305 | |||
XM_005250523.4 | XP_005250580.1 | 8 | 1289 | |||
XM_017012495.1 | XP_016867984.1 | 8 | 1289 | |||
GenBank mRNA | AB051486.1 | - | 8 | 1247 | ||
AF132734.1 | - | 2 | 261 | |||
AF380839.1 | - | 8 | 1302 | |||
AK022751.1 | - | 1 | 73 | |||
AK027688.1 | - | 8 | 1299 | |||
AK295404.1 | - | 8 | 1313 | |||
AK298191.1 | - | 3 | 287 | |||
AL831989.2 | - | 8 | 1300 | |||
AL834475.1 | - | 5 | 632 | |||
AW451395.1 | - | 3 | 312 | |||
BC020607.2 | - | 2 | 264 | |||
BC026174.1 | - | 8 | 1281 | |||
BC067263.1 | - | 8 | 1282 | 6183004 |