Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037126.1 | NP_001032203.1 | 3 | 378 | ||
NM_021807.3 | NP_068579.3 | 3 | 378 | |||
XM_005250523.4 | XP_005250580.1 | 3 | 362 | |||
XM_017012495.1 | XP_016867984.1 | 3 | 362 | |||
GenBank mRNA | AB051486.1 | - | 3 | 320 | ||
AF380839.1 | - | 3 | 375 | |||
AK027688.1 | - | 3 | 372 | |||
AK295404.1 | - | 3 | 386 | |||
BC026174.1 | - | 3 | 354 | |||
BC067263.1 | - | 3 | 355 | 6183004 |