Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOVL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242828.1 | NP_001229757.1 | 3 | 454 | ||
NM_001242830.1 | NP_001229759.1 | 3 | 454 | |||
NM_001301856.1 | NP_001288785.1 | 3 | 687 | |||
NM_021814.4 | NP_068586.1 | 3 | 454 | |||
GenBank mRNA | AB209798.1 | - | 3 | 202 | ||
AF231981.1 | - | 2 | 83 | |||
AF338241.1 | - | 3 | 427 | |||
AK074889.1 | - | 2 | 88 | |||
AK125098.1 | - | 3 | 531 | |||
AK299674.1 | - | 3 | 225 | |||
AK302948.1 | - | 3 | 225 | |||
AL136939.1 | - | 3 | 202 | |||
BC067123.1 | - | 3 | 203 | 6160926 | ||
DB225467.1 | - | 3 | 510 | |||
DC345924.1 | - | 3 | 455 |