Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIF3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136039.2 | NP_001129511.1 | 3 | 654 | ||
NM_001142355.1 | NP_001135827.1 | 3 | 603 | |||
NM_001142356.1 | NP_001135828.1 | 2 | 589 | |||
NM_021824.3 | NP_068596.2 | 3 | 806 | |||
XM_005246742.3 | XP_005246799.1 | 3 | 869 | |||
XM_011511582.1 | XP_011509884.1 | 3 | 790 | |||
XM_011511583.1 | XP_011509885.1 | 3 | 1534 | |||
XM_011511584.1 | XP_011509886.1 | 3 | 1352 | |||
XM_011511585.1 | XP_011509887.1 | 3 | 1301 | |||
XM_017004630.1 | XP_016860119.1 | 3 | 1051 | |||
GenBank mRNA | AB038949.1 | - | 3 | 767 | ||
AF060513.1 | - | 3 | 563 | |||
AF182416.1 | - | 3 | 728 | |||
AF283538.1 | - | 3 | 609 | |||
AK023378.1 | - | 3 | 752 | |||
AK225905.1 | - | 3 | 2129 | |||
AK314709.1 | - | 3 | 608 | |||
AL834430.1 | - | 2 | 429 | |||
AY251943.1 | - | 2 | 592 | |||
AY251944.1 | - | 2 | 592 | |||
BC007654.2 | - | 2 | 590 | 3346099 |