Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021942.5 | NP_068761.4 | 11 | 1390 | ||
NM_199053.2 | NP_951008.1 | 11 | 1390 | |||
XM_017008537.1 | XP_016864026.1 | 11 | 1390 | |||
XM_017008538.1 | XP_016864027.1 | 11 | 1390 | |||
GenBank mRNA | AK091146.1 | - | 6 | 614 | ||
AK096345.1 | - | 11 | 1364 | |||
AL136752.1 | - | 3 | 299 | |||
AL833451.1 | - | 10 | 1198 | |||
AL833571.1 | - | 11 | 1369 | |||
BC051724.1 | - | 11 | 1334 | 5265781 | ||
BC139745.1 | - | 4 | 390 | |||
BX538044.1 | - | 11 | 1344 | |||
BX647127.1 | - | 11 | 1361 | |||
BX648499.1 | - | 11 | 1365 | |||
CR533553.1 | - | 3 | 234 |