Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021942.5 | NP_068761.4 | 8 | 958 | ||
NM_199053.2 | NP_951008.1 | 8 | 958 | |||
XM_017008537.1 | XP_016864026.1 | 8 | 958 | |||
XM_017008538.1 | XP_016864027.1 | 8 | 958 | |||
XM_017008539.1 | XP_016864028.1 | 7 | 703 | |||
GenBank mRNA | AK091146.1 | - | 3 | 182 | ||
AK096345.1 | - | 8 | 932 | |||
AK315057.1 | - | 3 | 164 | |||
AL833451.1 | - | 7 | 766 | |||
AL833571.1 | - | 8 | 937 | |||
BC051724.1 | - | 8 | 902 | 5265781 | ||
BX538044.1 | - | 8 | 912 | |||
BX647127.1 | - | 8 | 929 | |||
BX648499.1 | - | 8 | 933 |