Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130706.2 | NP_001124178.1 | 3 | 493 | ||
NM_001130708.2 | NP_001124180.1 | 2 | 414 | |||
NM_001282970.1 | NP_001269899.1 | 2 | 660 | |||
NM_021944.3 | NP_068763.2 | 2 | 660 | |||
XM_005267971.1 | XP_005268028.1 | 2 | 562 | |||
XM_005267972.1 | XP_005268029.1 | 2 | 463 | |||
XM_011537059.1 | XP_011535361.1 | 2 | 646 | |||
XM_011537060.2 | XP_011535362.1 | 3 | 458 | |||
XM_011537062.1 | XP_011535364.1 | 2 | 1715 | |||
XM_017021569.1 | XP_016877058.1 | 3 | 4525 | |||
XM_017021570.1 | XP_016877059.1 | 3 | 3810 | |||
XM_017021571.1 | XP_016877060.1 | 4 | 3931 | |||
XM_017021572.1 | XP_016877061.1 | 3 | 3770 | |||
XM_017021573.1 | XP_016877062.1 | 3 | 667 | |||
XM_017021574.1 | XP_016877063.1 | 4 | 3725 | |||
GenBank mRNA | AK022216.1 | - | 2 | 602 | ||
AK023026.1 | - | 2 | 347 | |||
BC014299.2 | - | 2 | 579 | 3936989 | ||
BX161383.1 | - | 4 | 597 | |||
BX161417.1 | - | 3 | 420 | |||
DA891690.1 | - | 2 | 414 |