Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286455.1 | NP_001273384.1 | 7 | 937 | ||
NM_015482.1 | NP_056297.1 | 7 | 1396 | |||
NM_021945.6 | NP_068764.3 | 8 | 1182 | |||
NR_104448.1 | 8 | 990 | ||||
XM_011514801.2 | XP_011513103.1 | 8 | 1483 | |||
XM_017011180.1 | XP_016866669.1 | 8 | 1960 | |||
XM_017011181.1 | XP_016866670.1 | 6 | 709 | |||
XM_017011182.1 | XP_016866671.1 | 5 | 991 | |||
XM_017011184.1 | XP_016866673.1 | 5 | 646 | |||
XM_017011186.1 | XP_016866675.1 | 8 | 879 | |||
GenBank mRNA | AK172770.1 | - | 4 | 492 | ||
AK308943.1 | - | 7 | 880 | |||
BC038748.1 | - | 8 | 1153 | |||
BC128580.1 | - | 7 | 889 | 40128787 | ||
BC128581.1 | - | 8 | 946 |
Gene Symbol: LOC100422781 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096219.1 | - | 2 | 454 |