Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286455.1 | NP_001273384.1 | 8, 9 | 1131 | ||
NM_015482.1 | NP_056297.1 | 8, 9 | 1590 | |||
NM_021945.6 | NP_068764.3 | 9, 10 | 1376 | |||
NR_104448.1 | 9, 10 | 1184 | ||||
XM_011514801.2 | XP_011513103.1 | 9, 10 | 1677 | |||
XM_017011180.1 | XP_016866669.1 | 9, 10 | 2154 | |||
XM_017011181.1 | XP_016866670.1 | 7, 8 | 903 | |||
XM_017011182.1 | XP_016866671.1 | 6, 7 | 1185 | |||
XM_017011184.1 | XP_016866673.1 | 6, 7 | 840 | |||
XM_017011186.1 | XP_016866675.1 | 9, 10 | 1073 | |||
GenBank mRNA | AK172770.1 | - | 5, 6 | 686 | ||
AK308282.1 | - | 9, 10 | 1129 | |||
AK308943.1 | - | 8, 9 | 1074 | |||
BC038748.1 | - | 9, 10 | 1347 | |||
BC128580.1 | - | 8, 9 | 1083 | 40128787 | ||
BC128581.1 | - | 9, 10 | 1140 |