Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCORL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184772.2 | NP_001171701.1 | 4, 5 | 3649 | ||
NM_021946.4 | NP_068765.3 | 4, 5 | 3649 | |||
XM_005262452.4 | XP_005262509.2 | 6, 7 | 4065 | |||
XM_005262453.4 | XP_005262510.2 | 6, 7 | 3833 | |||
XM_005262454.3 | XP_005262511.2 | 5, 6 | 3777 | |||
XM_005262455.4 | XP_005262512.2 | 5, 6 | 3720 | |||
XM_005262456.4 | XP_005262513.2 | 6, 7 | 4065 | |||
XM_006724776.3 | XP_006724839.1 | 6, 7 | 3850 | |||
XM_006724777.3 | XP_006724840.1 | 5, 6 | 3745 | |||
XM_006724779.2 | XP_006724842.1 | 6, 7 | 3936 | |||
XM_017029721.1 | XP_016885210.1 | 6, 7 | 4096 | |||
XM_017029722.1 | XP_016885211.1 | 5, 6 | 4150 | |||
GenBank mRNA | AK074089.1 | - | 4, 5 | 3562 | ||
AL096777.1 | - | 2, 3 | 1911 |