Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022064.4 | NP_071347.2 | 32 | 3237 | ||
NR_135218.1 | 32 | 3477 | ||||
XM_006713285.1 | XP_006713348.1 | 32 | 3395 | |||
XM_011533995.1 | XP_011532297.1 | 32 | 3172 | |||
XM_017007018.1 | XP_016862507.1 | 32 | 3199 | |||
GenBank mRNA | AF370367.1 | - | 23 | 2391 | ||
AK022627.1 | - | 10 | 1063 | |||
AK026968.1 | - | 14 | 1491 | |||
AK090632.1 | - | 22 | 2279 | |||
AK096353.1 | - | 32 | 3151 | |||
AK126883.1 | - | 16 | 2944 | |||
AL136729.1 | - | 5 | 621 | |||
AL832034.1 | - | 32 | 3199 | |||
AL832138.1 | - | 31 | 3488 | |||
AL832344.1 | - | 32 | 3175 | |||
AY744152.1 | - | 31 | 3120 | |||
BC041145.1 | - | 17 | 1770 | |||
BC057392.1 | - | 19 | 1927 | |||
BC088801.1 | - | 14 | 1498 | |||
BC130632.1 | - | 32 | 3178 | 40146663 |