Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022064.4 | NP_071347.2 | 32 | 3270 | ||
NR_135218.1 | 32 | 3510 | ||||
XM_006713285.1 | XP_006713348.1 | 32 | 3428 | |||
XM_011533995.1 | XP_011532297.1 | 32 | 3205 | |||
XM_017007018.1 | XP_016862507.1 | 32 | 3232 | |||
GenBank mRNA | AF370367.1 | - | 23 | 2424 | ||
AK022627.1 | - | 10 | 1096 | |||
AK026968.1 | - | 14 | 1524 | |||
AK090632.1 | - | 22 | 2312 | |||
AK096353.1 | - | 32 | 3184 | |||
AK126883.1 | - | 16 | 2977 | |||
AL136729.1 | - | 5 | 654 | |||
AL832034.1 | - | 32 | 3232 | |||
AL832138.1 | - | 31 | 3521 | |||
AL832344.1 | - | 32 | 3208 | |||
AY744152.1 | - | 31 | 3153 | |||
BC041145.1 | - | 17 | 1803 | |||
BC057392.1 | - | 19 | 1960 | |||
BC088801.1 | - | 14 | 1531 | |||
BC130632.1 | - | 32 | 3211 | 40146663 | ||
CR627051.1 | - | 28 | 4842 |