Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022064.4 | NP_071347.2 | 33 | 3355 | ||
NR_135218.1 | 33 | 3595 | ||||
XM_006713285.1 | XP_006713348.1 | 33 | 3513 | |||
XM_011533995.1 | XP_011532297.1 | 33 | 3290 | |||
XM_017007018.1 | XP_016862507.1 | 33 | 3317 | |||
GenBank mRNA | AF370367.1 | - | 24 | 2509 | ||
AK022627.1 | - | 11 | 1181 | |||
AK026968.1 | - | 15 | 1609 | |||
AK090632.1 | - | 23 | 2397 | |||
AK096353.1 | - | 33 | 3269 | |||
AK126883.1 | - | 17 | 3062 | |||
AL136729.1 | - | 6 | 739 | |||
AL832034.1 | - | 33 | 3317 | |||
AL832138.1 | - | 32 | 3606 | |||
AL832344.1 | - | 33 | 3293 | |||
AY744152.1 | - | 32 | 3238 | |||
BC041145.1 | - | 18 | 1888 | |||
BC057392.1 | - | 20 | 2045 | |||
BC088801.1 | - | 15 | 1616 | |||
BC130632.1 | - | 33 | 3296 | 40146663 | ||
CR627051.1 | - | 29 | 4927 |