Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022066.3 | NP_071349.3 | 16 | 3062 | ||
XM_005257567.1 | XP_005257624.1 | 16 | 3049 | |||
XM_017024934.1 | XP_016880423.1 | 16 | 2974 | |||
XM_017024935.1 | XP_016880424.1 | 16 | 2971 | |||
GenBank mRNA | AB051521.1 | - | 16 | 3062 | ||
AK022940.1 | - | 3 | 560 | |||
AK024657.1 | - | 1 | 61 | |||
AK025999.1 | - | 8 | 1541 | |||
AK225412.1 | - | 8 | 1541 | |||
AL832432.1 | - | 9 | 1987 | |||
BC022237.1 | - | 3 | 580 | |||
BC025977.1 | - | 3 | 461 | |||
BC036820.1 | - | 11 | 2198 | |||
BC051868.2 | - | 8 | 1405 | 6162673 |