Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIPAS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193314.1 | NP_001180243.1 | 9, 10 | 1173 | ||
NM_001193315.1 | NP_001180244.1 | 9, 10 | 773 | |||
NM_001193316.1 | NP_001180245.1 | 8, 9 | 626 | |||
NM_001193317.1 | NP_001180246.1 | 9, 10 | 950 | |||
NM_022067.3 | NP_071350.2 | 10, 11 | 1044 | |||
XM_011537066.1 | XP_011535368.1 | 8, 9 | 671 | |||
XM_017021579.1 | XP_016877068.1 | 8, 9 | 805 | |||
XM_017021580.1 | XP_016877069.1 | 9, 10 | 764 | |||
XM_017021581.1 | XP_016877070.1 | 9, 10 | 898 | |||
GenBank mRNA | AK022769.1 | - | 9, 10 | 721 | ||
AK022925.1 | - | 9, 10 | 699 | |||
AK098254.1 | - | 9, 10 | 1134 | |||
AK222706.1 | - | 9, 10 | 719 | |||
AK297388.1 | - | 9, 10 | 875 | |||
AK298354.1 | - | 8, 9 | 572 | |||
BC015054.1 | - | 10, 11 | 965 | 3920459 | ||
CR457294.1 | - | 8, 9 | 627 |