Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM111A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142519.2 | NP_001135991.1 | 4, 5 | 579 | ||
NM_001142520.2 | NP_001135992.1 | 4, 5 | 602 | |||
NM_001142521.2 | NP_001135993.1 | 4, 5 | 415 | |||
NM_001312909.1 | NP_001299838.1 | 5, 6 | 817 | |||
NM_001312910.1 | NP_001299839.1 | 4, 5 | 641 | |||
NM_001312911.1 | NP_001299840.1 | 3, 4 | 406 | |||
NM_022074.3 | NP_071357.2 | 3, 4 | 313 | |||
NM_198847.2 | NP_942144.1 | 3, 4 | 274 | |||
GenBank mRNA | AB067482.1 | - | 1, 2 | 717 | ||
AK092061.1 | - | 3, 4 | 257 | |||
AK092237.1 | - | 4, 5 | 2907 | |||
AK291453.1 | - | 3, 4 | 279 | |||
BC071759.1 | - | 3, 4 | 273 | 6451741 | ||
BQ420366.1 | - | 3, 4 | 406 | |||
BQ948815.1 | - | 4, 5 | 502 | |||
CR747277.1 | - | 4, 5 | 427 | |||
CR749358.1 | - | 2, 3 | 920 | |||
DA502464.1 | - | 3, 4 | 313 | |||
DB218857.1 | - | 4, 5 | 480 |