Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM111A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142519.2 | NP_001135991.1 | 5 | 1177 | ||
NM_001142520.2 | NP_001135992.1 | 5 | 1200 | |||
NM_001142521.2 | NP_001135993.1 | 5 | 1013 | |||
NM_001312909.1 | NP_001299838.1 | 6 | 1415 | |||
NM_001312910.1 | NP_001299839.1 | 5 | 1239 | |||
NM_001312911.1 | NP_001299840.1 | 4 | 1004 | |||
NM_022074.3 | NP_071357.2 | 4 | 911 | |||
NM_198847.2 | NP_942144.1 | 4 | 872 | |||
GenBank mRNA | AB067482.1 | - | 2 | 1315 | ||
AK092061.1 | - | 4 | 855 | |||
AK092237.1 | - | 5 | 3505 | |||
AK092953.1 | - | 1 | 91 | |||
AK291453.1 | - | 4 | 877 | |||
BC013137.1 | - | 1 | 107 | |||
BC071759.1 | - | 4 | 871 | 6451741 | ||
CR749358.1 | - | 3 | 1518 |