Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022095.3 | NP_071378.1 | 24 | 3726 | ||
XM_005260504.4 | XP_005260561.1 | 24 | 4439 | |||
XM_017028012.1 | XP_016883501.1 | 23 | 3497 | |||
GenBank mRNA | AF395833.1 | - | 24 | 3703 | ||
AK026157.1 | - | 16 | 2241 | |||
AK074214.1 | - | 9 | 1361 | |||
AK122963.1 | - | 7 | 1917 | |||
AK296989.1 | - | 21 | 3297 | |||
BC041331.2 | - | 21 | 2941 | |||
BC142981.1 | - | 21 | 2941 |