Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCUR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031713.3 | NP_001026883.1 | 1, 2 | 537 | ||
XM_011514802.1 | XP_011513104.1 | 1, 2 | 540 | |||
GenBank mRNA | AK024611.1 | - | 1, 2 | 546 | ||
BC008234.1 | - | 1, 2 | 292 | |||
BC016850.1 | - | 1, 2 | 511 | 3886144 |