Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024504.3 | NP_078780.1 | 6 | 1558 | ||
XM_011517572.1 | XP_011515874.1 | 5 | 838 | |||
GenBank mRNA | AF319458.1 | - | 6 | 1558 | ||
AK022595.1 | - | 6 | 1558 | |||
BC052311.1 | - | 6 | 1471 | 6671984 | ||
BF770200.1 | - | 3 | 384 | |||
KJ463837.1 | - | 5 | 1356 |