Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313726.1 | NP_001300655.1 | 13 | 1472 | ||
NM_001313727.1 | NP_001300656.1 | 9 | 1030 | |||
NM_031418.3 | NP_113606.2 | 12 | 1380 | |||
XM_017018118.1 | XP_016873607.1 | 10 | 1008 | |||
XM_017018119.1 | XP_016873608.1 | 7 | 620 | |||
GenBank mRNA | AJ300461.1 | - | 12 | 2087 | ||
AK295143.1 | - | 9 | 943 | |||
AK295816.1 | - | 9 | 942 | |||
AK302659.1 | - | 12 | 1419 | |||
AK304781.1 | - | 13 | 1313 | |||
AK316018.1 | - | 9 | 942 |