Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033054.2 | NP_149043.2 | 15, 16 | 2082 | ||
XM_017012503.1 | XP_016867992.1 | 14, 15 | 1841 | |||
XM_017012504.1 | XP_016867993.1 | 15, 16 | 2077 | |||
GenBank mRNA | AB290179.1 | - | 15, 16 | 2042 | ||
AF380932.1 | - | 14 | 1842 | |||
AF380933.1 | - | 14 | 1842 | |||
AK074135.1 | - | 11 | 1855 | |||
AK131089.1 | - | 10, 11 | 4199 | |||
AL832302.1 | - | 14, 15 | 1837 | |||
BC113544.1 | - | 15, 16 | 2029 | 8322596 | ||
BC117241.1 | - | 15, 16 | 2029 | 40125792 |