Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PERP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022121.4 | NP_071404.2 | 3 | 661 | ||
GenBank mRNA | AF317550.1 | - | 3 | 550 | ||
AK074585.1 | - | 3 | 588 | |||
AK097958.1 | - | 3 | 575 | |||
AK314526.1 | - | 3 | 557 | |||
BC010163.2 | - | 3 | 537 | 3639866 |