Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PERP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022121.4 | NP_071404.2 | 3 | 738 | ||
GenBank mRNA | AF317550.1 | - | 3 | 627 | ||
AJ251830.1 | - | 3 | 592 | |||
AK074585.1 | - | 3 | 665 | |||
AK075082.1 | - | 3 | 634 | |||
AK093516.1 | - | 4 | 580 | |||
AK097387.1 | - | 3 | 547 | |||
AK097958.1 | - | 3 | 652 | |||
AK225907.1 | - | 3 | 631 | |||
AK314526.1 | - | 3 | 634 | |||
AY157578.1 | - | 3 | 628 | |||
BC010163.2 | - | 3 | 614 | 3639866 | ||
DR422553.1 | - | 1 | 52 |