Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMSN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256370.1 | NP_001243299.1 | 4 | 598 | ||
NM_001286523.1 | NP_001273452.1 | 4 | 442 | |||
NM_022136.4 | NP_071419.3 | 3 | 318 | |||
XM_011529684.2 | XP_011527986.1 | 4 | 358 | |||
XM_011529686.1 | XP_011527988.1 | 3 | 315 | |||
XM_017028427.1 | XP_016883916.1 | 3 | 315 | |||
GenBank mRNA | AF218085.2 | - | 4 | 598 | ||
AF222927.1 | - | 3 | 301 | |||
AK125144.1 | - | 3 | 301 | |||
BC029112.1 | - | 3 | 292 | 4343284 | ||
DA819791.1 | - | 3 | 318 | |||
HG380513.1 | - | 4 | 348 | |||
HG380514.1 | - | 4 | 417 |