Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166412.1 | NP_001159884.1 | 7 | 809 | ||
NM_022138.2 | NP_071421.1 | 7 | 842 | |||
XM_011536065.1 | XP_011534367.1 | 7 | 840 | |||
XM_011536066.1 | XP_011534368.1 | 7 | 807 | |||
GenBank mRNA | AB014730.1 | - | 7 | 609 | ||
AB014737.1 | - | 7 | 642 | |||
AF173892.1 | - | 6 | 464 | |||
AJ249902.1 | - | 2 | 88 | |||
AJ420521.1 | - | 7 | 789 | |||
AK056700.1 | - | 7 | 736 | |||
AK314613.1 | - | 7 | 776 | |||
AL832303.1 | - | 7 | 744 | |||
BC028420.1 | - | 7 | 812 | 4826774 | ||
BC047583.1 | - | 7 | 781 | 4826950 |