Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARVG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137605.2 | NP_001131077.1 | 12 | 1159 | ||
NM_022141.6 | NP_071424.1 | 12 | 1219 | |||
XM_005261702.3 | XP_005261759.1 | 12 | 946 | |||
XM_011530302.2 | XP_011528604.1 | 11 | 3711 | |||
XM_017028907.1 | XP_016884396.1 | 12 | 2157 | |||
XM_017028908.1 | XP_016884397.1 | 12 | 2177 | |||
GenBank mRNA | AF237772.1 | - | 12 | 1013 | ||
AK225929.1 | - | 13 | 1109 | |||
AL355092.1 | - | 10 | 926 | |||
AL590887.1 | - | 10 | 735 | |||
BC034406.1 | - | 12 | 1140 | 5178529 | ||
BX648942.1 | - | 7 | 4076 | |||
CA440742.1 | - | 3 | 430 | |||
CR456480.2 | - | 11 | 790 |