Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022144.2 | NP_071427.2 | 5, 6 | 794 | ||
GenBank mRNA | AB055421.1 | - | 5, 6 | 574 | ||
AF191770.1 | - | 5, 6 | 754 | |||
AF234259.1 | - | 5, 6 | 635 | |||
AF291656.1 | - | 5, 6 | 640 | |||
AY358706.1 | - | 5, 6 | 626 |