Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142505.1 | NP_001135977.1 | 12 | 1461 | ||
NM_022169.4 | NP_071452.2 | 12 | 1685 | |||
XM_011542952.2 | XP_011541254.1 | 11 | 1713 | |||
XM_017018121.1 | XP_016873610.1 | 12 | 1416 | |||
GenBank mRNA | AJ300465.1 | - | 11 | 1348 | ||
AJ308237.1 | - | 11 | 1360 | |||
AJ308248.1 | - | 1 | 21 | |||
AK289830.1 | - | 12 | 1461 | |||
AK290035.1 | - | 12 | 1622 | |||
AK308673.1 | - | 9 | 1255 | |||
AL137563.1 | - | 6 | 661 | |||
BC041091.1 | - | 12 | 1685 | 5763981 |