Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PJA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032396.2 | NP_001027568.1 | 2 | 1849 | ||
NM_022368.4 | NP_071763.2 | 3 | 1611 | |||
NM_145119.3 | NP_660095.1 | 2 | 2175 | |||
XM_005262292.1 | XP_005262349.1 | 2 | 1837 | |||
XM_011531011.2 | XP_011529313.1 | 2 | 1982 | |||
GenBank mRNA | AF262024.1 | - | 1 | 2104 | ||
AF264620.1 | - | 3 | 1526 | |||
AK021892.1 | - | 1 | 954 | |||
AK096611.1 | - | 2 | 2174 | |||
AK226171.1 | - | 2 | 1756 | |||
AK290543.1 | - | 2 | 2075 | |||
AK293266.1 | - | 3 | 1655 | |||
AK294632.1 | - | 3 | 1688 | |||
BC039149.1 | - | 1 | 1181 | |||
BC048323.1 | - | 1 | 1021 | |||
BC058327.1 | - | 1 | 1161 | |||
BC075803.1 | - | 2 | 2082 | 3913310 | ||
BC105051.1 | - | 1 | 1864 | 8144054 | ||
BC105053.1 | - | 1 | 1864 | 8144056 |