Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142617.1 | NP_001136089.1 | 13 | 1361 | ||
NM_001142618.1 | NP_001136090.1 | 13 | 1343 | |||
NM_001142619.1 | NP_001136091.1 | 13 | 1334 | |||
NM_001199040.1 | NP_001185969.1 | 13 | 1477 | |||
NM_001199041.1 | NP_001185970.1 | 13 | 1488 | |||
NM_001199042.1 | NP_001185971.1 | 13 | 1592 | |||
NM_022369.3 | NP_071764.3 | 13 | 1297 | |||
XM_011521883.1 | XP_011520185.1 | 13 | 1263 | |||
XM_011521884.1 | XP_011520186.1 | 13 | 1404 | |||
XM_017022478.1 | XP_016877967.1 | 13 | 1212 | |||
XM_017022479.1 | XP_016877968.1 | 13 | 1303 | |||
XM_017022480.1 | XP_016877969.1 | 13 | 1327 | |||
GenBank mRNA | AF352728.1 | - | 13 | 1163 | ||
AF352729.1 | - | 13 | 1273 | |||
AF370419.2 | - | 13 | 1269 | |||
AK022603.1 | - | 9 | 847 | |||
AK056125.1 | - | 13 | 1358 | |||
AK091152.1 | - | 13 | 1387 | |||
AK092227.1 | - | 8 | 824 | |||
AK291966.1 | - | 13 | 1233 | |||
AK298921.1 | - | 13 | 1232 | |||
AK298928.1 | - | 11 | 901 | |||
AK299191.1 | - | 13 | 1477 | |||
AK302932.1 | - | 13 | 1488 | |||
AK309676.1 | - | 13 | 1572 | |||
AK310878.1 | - | 12 | 1350 | |||
AY358748.1 | - | 13 | 1273 | |||
AY359089.1 | - | 13 | 1163 | |||
BC025256.1 | - | 13 | 1269 | 4892821 | ||
BX537413.1 | - | 13 | 1239 |