Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142617.1 | NP_001136089.1 | 14 | 1502 | ||
NM_001142618.1 | NP_001136090.1 | 14 | 1484 | |||
NM_001142619.1 | NP_001136091.1 | 14 | 1475 | |||
NM_001199040.1 | NP_001185969.1 | 14 | 1618 | |||
NM_001199041.1 | NP_001185970.1 | 14 | 1629 | |||
NM_001199042.1 | NP_001185971.1 | 14 | 1733 | |||
NM_022369.3 | NP_071764.3 | 14 | 1438 | |||
XM_011521883.1 | XP_011520185.1 | 14 | 1404 | |||
XM_011521884.1 | XP_011520186.1 | 14 | 1545 | |||
XM_017022478.1 | XP_016877967.1 | 14 | 1353 | |||
XM_017022479.1 | XP_016877968.1 | 14 | 1444 | |||
XM_017022480.1 | XP_016877969.1 | 14 | 1468 | |||
GenBank mRNA | AF352728.1 | - | 14 | 1304 | ||
AF352729.1 | - | 14 | 1414 | |||
AF370419.2 | - | 14 | 1410 | |||
AK022603.1 | - | 10 | 988 | |||
AK056125.1 | - | 14 | 1499 | |||
AK091152.1 | - | 14 | 1528 | |||
AK092227.1 | - | 9 | 965 | |||
AK291966.1 | - | 14 | 1374 | |||
AK298921.1 | - | 14 | 1373 | |||
AK298928.1 | - | 12 | 1042 | |||
AK302932.1 | - | 14 | 1629 | |||
AK309676.1 | - | 14 | 1713 | |||
AK310878.1 | - | 13 | 1491 | |||
AY358748.1 | - | 14 | 1414 | |||
AY359089.1 | - | 14 | 1304 | |||
BC025256.1 | - | 14 | 1410 | 4892821 | ||
BX537413.1 | - | 14 | 1380 |