Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022436.2 | NP_071881.1 | 1 | 179 | ||
XM_005264480.3 | XP_005264537.1 | 1 | 200 | |||
XM_006712073.3 | XP_006712136.1 | 1 | 200 | |||
XM_006712074.3 | XP_006712137.1 | 1 | 200 | |||
XM_011533024.2 | XP_011531326.1 | 1 | 200 | |||
XM_011533025.2 | XP_011531327.1 | 1 | 124 | |||
XM_011533026.2 | XP_011531328.1 | 1 | 200 | |||
XM_011533027.2 | XP_011531329.1 | 1 | 124 | |||
GenBank mRNA | AF312715.2 | - | 1 | 179 | ||
AF320293.1 | - | 1 | 145 | |||
AK091997.1 | - | 1 | 225 | |||
BC111541.1 | - | 1 | 110 |