Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022449.3 | NP_071894.1 | 2 | 789 | ||
NR_033308.1 | 2 | 789 | ||||
XM_017004693.1 | XP_016860182.1 | 2 | 738 | |||
GenBank mRNA | AK022600.1 | - | 2 | 772 | ||
AK225423.1 | - | 2 | 738 | |||
AL136645.1 | - | 2 | 396 | |||
BC007907.2 | - | 2 | 151 | |||
BC009827.2 | - | 2 | 382 | |||
BC050426.1 | - | 2 | 387 | 5761178 | ||
CR457282.1 | - | 1 | 126 | |||
CR533454.1 | - | 1 | 126 |