Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOC3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022451.10 | NP_071896.8 | 17 | 2008 | ||
XM_017016544.1 | XP_016872033.1 | 15 | 2061 | |||
GenBank mRNA | AB077992.1 | - | 17 | 1895 | ||
AK022882.1 | - | 9 | 919 | |||
AK026953.1 | - | 7 | 619 | |||
AK091246.1 | - | 17 | 1961 | |||
AK292520.1 | - | 17 | 1990 | |||
AK302028.1 | - | 11 | 1093 | |||
BC049850.1 | - | 17 | 1958 | 5549517 |