Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022458.3 | NP_071903.2 | 11 | 1109 | ||
XM_005249555.2 | XP_005249612.1 | 11 | 1084 | |||
XM_017012507.1 | XP_016867996.1 | 12 | 1207 | |||
XM_017012508.1 | XP_016867997.1 | 12 | 1165 | |||
XM_017012509.1 | XP_016867998.1 | 12 | 1207 | |||
XM_017012510.1 | XP_016867999.1 | 10 | 811 | |||
XM_017012511.1 | XP_016868000.1 | 10 | 940 | |||
XM_017012512.1 | XP_016868001.1 | 10 | 871 | |||
XM_017012513.1 | XP_016868002.1 | 11 | 889 | |||
XM_017012514.1 | XP_016868003.1 | 12 | 1178 | |||
XM_017012516.1 | XP_016868005.1 | 10 | 743 | |||
XM_017012517.1 | XP_016868006.1 | 12 | 1162 | |||
XM_017012518.1 | XP_016868007.1 | 13 | 1260 | |||
GenBank mRNA | AF402318.1 | - | 11 | 920 | ||
AK299753.1 | - | 10 | 1007 | |||
AL833934.1 | - | 8 | 655 | |||
AL834394.1 | - | 12 | 1011 | |||
BC017663.1 | - | 11 | 1048 | 3883022 | ||
BC063649.1 | - | 13 | 1256 |