Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGAP9 SIRNA Assays ›
Gene Symbol:
ARHGAP9
Gene Name:
Rho GTPase activating protein 9
Gene Aliases:
10C, RGL1
Chromosome Location:
Chr.12: 57472255 - 57488814 on Build GRCh38
UniGene ID:
Hs.437126
Species:
Human
siRNA ID
HSS127537
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGAP9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080156.2
NP_001073625.1
13, 14
1664
NM_001080157.1
NP_001073626.1
14, 15
2019
NM_001319850.1
NP_001306779.1
18, 19
2247
NM_001319851.1
NP_001306780.1
12, 13
1940
NM_001319852.1
NP_001306781.1
13, 14
1721
NM_032496.3
NP_115885.2
15, 16
1945
XM_005269083.2
XP_005269140.1
13, 14
1386
XM_005269084.2
XP_005269141.1
12, 13
1319
XM_005269085.2
XP_005269142.1
12, 13
1293
XM_011538656.2
XP_011536958.1
17, 18
1982
XM_011538657.2
XP_011536959.1
14, 15
1823
XM_011538658.2
XP_011536960.1
14, 15
1765
XM_011538659.2
XP_011536961.1
17, 18
1982
XM_017019800.1
XP_016875289.1
14, 15
2259
XM_017019801.1
XP_016875290.1
13, 14
1329
XM_017019802.1
XP_016875291.1
12, 13
1262
XM_017019803.1
XP_016875292.1
13, 14
1329
XM_017019804.1
XP_016875293.1
12, 13
1262
GenBank mRNA
AB030239.1
-
13, 14
1416
AB051853.1
-
15, 16
1923
AK057573.1
-
12, 13
1593
AK092763.1
-
14, 15
2010
AK131415.1
-
18, 19
2174
AK301095.1
-
13, 14
1316
AL713803.1
-
8, 9
1008
BC006107.1
-
18, 19
2247
3010091
BC011820.1
-
13, 14
1390
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
GTPase activator activity
protein binding
phosphatidylinositol-3,4,5-trisphosphate binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline