Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205319.1 | NP_001192248.1 | 8 | 997 | ||
NM_022463.4 | NP_071908.2 | 8 | 1340 | |||
XM_005256756.4 | XP_005256813.1 | 7 | 1166 | |||
XM_005256758.3 | XP_005256815.1 | 8 | 1669 | |||
GenBank mRNA | AF086523.1 | - | 2 | 168 | ||
AK022676.1 | - | 6 | 769 | |||
AK027451.1 | - | 7 | 893 | |||
AK297990.1 | - | 4 | 469 | |||
AY189690.1 | - | 7 | 880 | |||
BC009327.2 | - | 8 | 934 | |||
BC063828.1 | - | 5 | 595 | |||
BC073845.1 | - | 6 | 750 |