Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037633.1 | NP_001032722.1 | 8 | 936 | ||
NM_022464.4 | NP_071909.1 | 7 | 848 | |||
XM_011543570.2 | XP_011541872.1 | 9 | 973 | |||
GenBank mRNA | AF547994.1 | - | 7 | 767 | ||
AJ299442.1 | - | 7 | 826 | |||
AK074624.1 | - | 7 | 840 | |||
AK075177.1 | - | 7 | 777 | |||
AY358950.1 | - | 7 | 794 | |||
BC011568.1 | - | 7 | 816 | 4640182 | ||
DQ059315.1 | - | 2 | 148 | |||
DQ173771.1 | - | 2 | 136 | |||
DQ173772.1 | - | 2 | 148 | |||
KU178743.1 | - | 6 | 730 | |||
KU178744.1 | - | 5 | 538 |