Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284306.1 | NP_001271235.1 | 14 | 3199 | ||
NM_001284307.1 | NP_001271236.1 | 16 | 3214 | |||
NM_022473.2 | NP_071918.1 | 15 | 5642 | |||
XM_005254591.2 | XP_005254648.1 | 18 | 5549 | |||
XM_005254592.2 | XP_005254649.1 | 18 | 5288 | |||
XM_017022483.1 | XP_016877972.1 | 17 | 5324 | |||
XM_017022484.1 | XP_016877973.1 | 15 | 5069 | |||
XM_017022485.1 | XP_016877974.1 | 16 | 2948 | |||
GenBank mRNA | AF205632.1 | - | 15 | 5615 | ||
AK024726.1 | - | 4 | 471 | |||
AK127741.1 | - | 13 | 2181 | |||
AK226132.1 | - | 16 | 3122 | |||
AK302735.1 | - | 17 | 3096 | |||
AL831983.1 | - | 14 | 3145 | |||
AL832014.1 | - | 14 | 3423 | |||
AL832352.1 | - | 14 | 3642 | |||
AL832367.1 | - | 13 | 2093 | |||
AL832455.2 | - | 14 | 3555 | |||
AL833301.1 | - | 14 | 3653 | |||
BX648629.1 | - | 14 | 4747 |