Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284306.1 | NP_001271235.1 | 16, 17 | 3376 | ||
NM_001284307.1 | NP_001271236.1 | 18, 19 | 3391 | |||
NM_022473.2 | NP_071918.1 | 17, 18 | 5819 | |||
XM_005254591.2 | XP_005254648.1 | 20, 21 | 5726 | |||
XM_005254592.2 | XP_005254649.1 | 20, 21 | 5465 | |||
XM_017022483.1 | XP_016877972.1 | 19, 20 | 5501 | |||
XM_017022484.1 | XP_016877973.1 | 17, 18 | 5246 | |||
XM_017022485.1 | XP_016877974.1 | 18, 19 | 3125 | |||
GenBank mRNA | AF205632.1 | - | 17, 18 | 5792 | ||
AK024726.1 | - | 6, 7 | 648 | |||
AK127741.1 | - | 15, 16 | 2358 | |||
AK226132.1 | - | 18, 19 | 3299 | |||
AK302735.1 | - | 19, 20 | 3273 | |||
AL831983.1 | - | 16, 17 | 3322 | |||
AL832014.1 | - | 16, 17 | 3600 | |||
AL832352.1 | - | 16, 17 | 3819 | |||
AL832367.1 | - | 15, 16 | 2270 | |||
AL832455.2 | - | 16, 17 | 3732 | |||
AL833301.1 | - | 16, 17 | 3830 | |||
BX648629.1 | - | 16, 17 | 4924 |